Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
123-38-6
C3H6O
58.08
NBBJYMSMWIIQGU-UHFFFAOYSA-N
propionaldehyde,propanaldehyde,propaldehyde,propional,propionic aldehyde,n-propanal,methylacetaldehyde,propylaldehyde,propylic aldehyde,aldehyde propionique
527
CHEBI:17153
propanal
CCC=O
123-38-6
C3H6O
58.08
NBBJYMSMWIIQGU-UHFFFAOYSA-N
propionaldehyde,propanaldehyde,propaldehyde,propional,propionic aldehyde,n-propanal,methylacetaldehyde,propylaldehyde,propylic aldehyde,aldehyde propionique
527
CHEBI:17153
propanal
CCC=O
Quantity 10L
Packaging Plastic drum
Viscosity 0.6 mPa.s (20°C)
Density 0.7980g/mL
Color Colorless
Specific Gravity 0.798
Infrared Spectrum Authentic
Melting Point -81.0°C
Refractive Index 1.3610 to 1.3650
Boiling Point 48.0°C
Flash Point −40°C
Assay Percent Range 97%
Linear Formula C2H5CHO
Beilstein 01,629
Merck Index 15,7937
Solubility Information Solubility in water: 540g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 58.08
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Propionaldehyde
Show More Show Less