Alfa Aesar™ Propionamide, 98%

Manufacturer: Alfa Aesar™ A1052122

Quantity 100g

 View more versions of this product

Catalog No. AAA1052122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-05-0
    Molecular Formula CH3CH2CONH2
    Formula Weight 73.1
    MDL Number MFCD00008039
    Merck Index 14,7824
    Chemical Name or Material Propionamide

    Specifications

    Formula Weight 73.1
    Molecular Formula CH3CH2CONH2
    CAS 79-05-0
    TSCA TSCA
    MDL Number MFCD00008039
    Merck Index 14,7824
    Boiling Point 213°C
    Melting Point 76° to 79°C
    Quantity 100g
    RTECS Number UE2975000
    Chemical Name or Material Propionamide
    Assay 98%

    Description

    Applications
    Propionamide was used as adsorbent in the determination of adsorption isotherms of acetamide and propionamide on multi-wall carbon nanotube1. It was used in a robust screening method to study biotransformations using (+)-γ-lactamase enzyme. Propionamide on γ-irradiation reacts with sulfur dioxide and this reaction has been studied by ESR spectroscopy, gas absorption measurements and X-ray diffraction3

    Solubility
    Soluble in water, chloroform, ethanol, diethyl ether.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids, bases, strong reducing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit