Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Propionamide, 98%

Manufacturer:  Alfa Aesar™ A1052136

Quantity 500g

 View more versions of this product

Catalog No. AAA1052136


  Check Availability
Add to cart

Specifications

Description

Applications
Propionamide was used as adsorbent in the determination of adsorption isotherms of acetamide and propionamide on multi-wall carbon nanotube1. It was used in a robust screening method to study biotransformations using (+)-γ-lactamase enzyme. Propionamide on γ-irradiation reacts with sulfur dioxide and this reaction has been studied by ESR spectroscopy, gas absorption measurements and X-ray diffraction3

Solubility
Soluble in water, chloroform, ethanol, diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids, bases, strong reducing agents.
Description & Specifications

Chemical Identifiers

79-05-0
73.1
14,7824
CH3CH2CONH2
MFCD00008039
Propionamide

Specifications

79-05-0
73.1
14,7824
76° to 79°C
Propionamide
CH3CH2CONH2
MFCD00008039
213°C
500g
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : UE2975000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit