Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Propionanilide, 98%

Manufacturer:  Alfa Aesar™ H3183206

Quantity 5g

 View more versions of this product

Catalog No. AAH3183206


  Check Availability
Add to cart

Specifications

Description

Applications
The propionanilide derivatives are the first developed non-steroidal antiandrogens and include drugs such as flutamide, hydroxyflutamide, nilutamide and bicalutamide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

620-71-3
149.19
N-Phenylpropionamide; N-Propionylaniline
C9H11NO
MFCD00092530
Propionanilide

Specifications

620-71-3
Odorless
MFCD00092530
222°C
5g
98%
C9H11NO
149.19
N-Phenylpropionamide; N-Propionylaniline
104° to 107°C
Propionanilide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

RTECSNumber : UE4600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit