Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic acid, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
79-09-4
C3H6O2
74.08
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propionic acid,ethylformic acid,methylacetic acid,carboxyethane,ethanecarboxylic acid,pseudoacetic acid,metacetonic acid,monoprop,luprosil,prozoin
1032
CHEBI:30768
propanoic acid
CCC(=O)O
79-09-4
C3H6O2
74.08
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propionic acid,ethylformic acid,methylacetic acid,carboxyethane,ethanecarboxylic acid,pseudoacetic acid,metacetonic acid,monoprop,luprosil,prozoin
1032
CHEBI:30768
propanoic acid
CCC(=O)O
Viscosity 1.02 mPa.s (25°C)
Boiling Point 141.0°C
Quantity 100mL
Flash Point 51°C
Specific Gravity 0.99
Packaging Glass bottle
Density 0.9900g/mL
Refractive Index 1.3850 to 1.3870
Melting Point -22.0°C
Linear Formula C2H5CO2H
Merck Index 15,7939
Solubility Information Solubility in water: soluble.
Formula Weight 74.08
Grade ACS Reagent
Chemical Name or Material Propionic acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 201-176-3