Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic Acid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

79-09-4
C3H6O2
74.079
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
CHEBI:30768

A grade of chemicals for laboratory use

CAS 79-09-4
Molecular Formula C3H6O2
Packaging Narrow Mouth Amber Glass Bottle
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILES CCC(=O)O
IUPAC Name propanoic acid
Molecular Weight (g/mol) 74.079
PubChem CID 1032
ChEBI CHEBI:30768
Formula Weight 74.08
Flash Point 52.00°C
Quantity 4L
Specific Gravity 1L = 0.99kg
Chemical Name or Material Propionic Acid
Show More Show Less