Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic Acid, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
79-09-4
C3H6O2
74.08
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
CCC(O)=O
79-09-4
C3H6O2
74.08
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
CCC(O)=O

P1326, 79-09-4, C3H6O2

Spectrum™ Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 99.5 to 100.5%
Lead (Pb) 2 mg/kg
Packaging Amber Glass Bottle
Specific Gravity 0.993-0.997
Water 0.0015
Quantity 4 x 4 L