Learn More
Propionic Acid, NF, 99.5-100.5%, Spectrum™
P1464, 79-09-4, C3H6O2
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner P14644LTGLCS4
 View more versions of this product
Catalog No. 18-605-885
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Spectrum™ Propionic Acid, NF is used as a preservative in pharmaceutical formulations. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
79-09-4
99.5 to 100.5%
4 x 4 L
1
Amber Glass Bottle