Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
500g
2.5kg
Packaging:
Glass bottle
79-03-8
C3H5ClO
92.52
RZWZRACFZGVKFM-UHFFFAOYSA-N
propionyl chloride,propionic acid chloride,propionic chloride,propionylchloride,unii-mb6vl5omb9,mb6vl5omb9,propanoylchloride,propionyl cloride,propanoic chloride,propionyl-chloride
62324
propanoyl chloride
CCC(=O)Cl
79-03-8
C3H5ClO
92.52
RZWZRACFZGVKFM-UHFFFAOYSA-N
propionyl chloride,propionic acid chloride,propionic chloride,propionylchloride,unii-mb6vl5omb9,mb6vl5omb9,propanoylchloride,propionyl cloride,propanoic chloride,propionyl-chloride
62324
propanoyl chloride
CCC(=O)Cl
Boiling Point 77.0°C to 79.0°C
Viscosity 0.48 mPa.s (20°C)
Refractive Index 1.4020 to 1.4050
Flash Point 11°C
Quantity 500g
Infrared Spectrum Authentic
Melting Point -94.0°C
Specific Gravity 1.06
Color Undesignated
Density 1.0600g/mL
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Linear Formula C2H5COCl
Beilstein 02,243
Merck Index 15,7942
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol
Formula Weight 92.52
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Propionyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Highly flammable liquid and vapor.
Harmful if swallowed.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 201-170-