Propionyl chloride, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar A1094036

Quantity 500g

 View more versions of this product

Catalog No. AAA1094036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 79-03-8
    Molecular Formula C3H5ClO
    Linear Formula CH3CH2COCl
    Formula Weight 92.53
    MDL Number MFCD00000745
    UN Number UN1815
    Beilstein 385632
    Merck Index 14,7828
    Chemical Name or Material Propionyl chloride

    Specifications

    Formula Weight 92.53
    Quantity 500g
    TSCA TSCA
    Molecular Formula C3H5ClO
    Linear Formula CH3CH2COCl
    Odor Unpleasant
    MDL Number MFCD00000745
    CAS 79-03-8
    UN Number UN1815
    Beilstein 385632
    Merck Index 14,7828
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314;
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Boiling Point 77° to 79°C
    Melting Point −94°C
    Flash Point 11°C (51°F)
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPIONYL CHLORIDE
    RTECS Number UC3926000
    Chemical Name or Material Propionyl chloride
    Assay 98%

    Description

    Applications
    Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.

    Notes
    Moisture sensitive. Incompatible with alcohols, oxidizing agents, strong bases and water.

    GHS H Statement
    H225-H314;
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapours/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit