Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propionyl-L-carnitine, ≥94.0% (HPLC), MilliporeSigma™ Supelco™
$149.00 - $483.00
Specifications
CAS | 20064-19-1 |
---|---|
Type | Analytical Standard |
Linear Formula | C10H19 NO4 |
MDL Number | MFCD00870024 |
Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium inner salt; L-Carnitine propionyl ester; C3-Carnitine; Propanoyl-L-carnitine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-3451
|
MilliporeSigma Supelco
9127510MG |
10 mg |
Each for $149.00
|
|
|||||
11-101-3452
|
MilliporeSigma Supelco
9127550MG |
50 mg |
Each for $483.00
|
|
|||||
Specifications
20064-19-1 | |
C10H19 NO4 | |
(2 R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium inner salt; L-Carnitine propionyl ester; C3-Carnitine; Propanoyl-L-carnitine | |
≥94.0% (HPLC) |
Analytical Standard | |
MFCD00870024 | |
217.26 | |
Analytical Standard |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title