Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Propionylcarnitine chloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
17298-37-2
C10H20ClNO4
253.723
KTFMPDDJYRFWQE-UHFFFAOYSA-N
+/--propionylcarnitine chloride,propionyl-l-carnitine,propionyllevocarnitine hydrochloride,3-carboxy-2-propanoyloxy propyl trimethylazanium chloride,a+/--propionylcarnitine chloride,3-carboxy-2-1-oxopropoxy propyl-trimethylammonium chloride,trimethyl-4-oxidanyl-4-oxidanylidene-2-propanoyloxy-butyl azanium chloride
13020033
(3-carboxy-2-propanoyloxypropyl)-trimethylazanium;chloride
CCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]
This item is not returnable. View return policy
17298-37-2
C10H20ClNO4
253.723
KTFMPDDJYRFWQE-UHFFFAOYSA-N
+/--propionylcarnitine chloride,propionyl-l-carnitine,propionyllevocarnitine hydrochloride,3-carboxy-2-propanoyloxy propyl trimethylazanium chloride,a+/--propionylcarnitine chloride,3-carboxy-2-1-oxopropoxy propyl-trimethylammonium chloride,trimethyl-4-oxidanyl-4-oxidanylidene-2-propanoyloxy-butyl azanium chloride
13020033
(3-carboxy-2-propanoyloxypropyl)-trimethylazanium;chloride
CCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Intermediate in lipid metabolism

Homolog of acetylcarnitine chloride (Cat. No. 0355). Acylcarnitines are important intermediates in lipid metabolism.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 253.73
Percent Purity >95%
Chemical Name or Material (±)-Propionylcarnitine chloride

Recommended Storage : Store at -20°C