Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propiophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
2mL
250mL
1L
2.5L
Packaging:
Glass bottle
93-55-0
C9H10O
134.18
KRIOVPPHQSLHCZ-UHFFFAOYSA-N
propiophenone,ethyl phenyl ketone,propionylbenzene,1-phenyl-1-propanone,1-propanone, 1-phenyl,phenyl ethyl ketone,ketone, ethyl phenyl,propionphenone,usaf ek-1235,1-phenyl-propan-1-one
7148
CHEBI:425902
1-phenylpropan-1-one
CCC(=O)C1=CC=CC=C1
93-55-0
C9H10O
134.18
KRIOVPPHQSLHCZ-UHFFFAOYSA-N
propiophenone,ethyl phenyl ketone,propionylbenzene,1-phenyl-1-propanone,1-propanone, 1-phenyl,phenyl ethyl ketone,ketone, ethyl phenyl,propionphenone,usaf ek-1235,1-phenyl-propan-1-one
7148
CHEBI:425902
1-phenylpropan-1-one
CCC(=O)C1=CC=CC=C1
Specific Gravity 1
Flash Point 87°C
Packaging Glass bottle
Refractive Index 1.5248 to 1.5268
Infrared Spectrum Authentic
Density 1.0000g/mL
Boiling Point 218.0°C
Melting Point 18.0°C
Color Colorless to Yellow
Quantity 2mL
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5COC2H5
Beilstein 07,II,231
Merck Index 15,7944
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with methanol, ether, benzene and toluene, insoluble in glycerol, ethylene glycol and, propylene glycol
Formula Weight 134.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Propiophenone
Show More Show Less

EINECSNumber : 202-257-6