Propper™ Indicator Tape
Placed on the outside of bag or instrument wrap to indicate when sterilization is complete
Manufacturer: Moore Medical Corp 17409
 View more versions of this product
Catalog No. 19-898-065
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Steam Sterilization
    For Use With (Equipment) Steam sterilizer
    Autoclavable Yes
    Product Type Indicator Tape
    Quantity Each
    Size (English) 0.75 x 2160 in.
    Size (Metric) 1.90 x 5486.4cm
  • Description & Specifications
Related Products
Product Content Feedback