Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propper Seracult™ Plus Single Slide Fecal Occult Blood Test

Click to view available options
Quantity:
100 Pk.
1000 Pk.
20 Pk.
Type:
Developer
Single Slide
Description
- Seracult™ Plus Fecal Occult Blood Test is a highly sensitive qualitative aid in the diagnosis of colorectal cancer.
- Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
- Seracult™ Plus was designed in mind for patients who are at an increased risk of disease.
- Seracult™ Plus Single Slides are ideal for testing inpatient or single samples at home.
- Seracult™ Plus is available in a variety of configurations to meet all of your needs.
- Results are ready instantly.
- Seracult guaiac paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
- Mailing kits of in-office slides.
- 28-day sample stability.
- CLIA Waived.
Specifications
Specifications
Clia Complexity | Waived |
Detectable Analytes | Hemoglobin |
Sample Type | Stool |
Type | Developer |
For Use With (Application) | Fecal Occult Blood Detection |
Includes | 20 x 15 mL Bottles of Developer |
Quantity | 20 Pk. |
Test Time | 30 to 60 sec. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction