Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propranolol Hydrochloride, USP, 98-101.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
318-98-9
C16H22ClNO2
295.81
ZMRUPTIKESYGQW-UHFFFAOYNA-N
hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
318-98-9
C16H22ClNO2
295.81
ZMRUPTIKESYGQW-UHFFFAOYNA-N
hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1

PR140, 318-98-9, C16H21NO2•HCl

Spectrum™ Propranolol Hydrochloride, USP is a sympatholytic nonselective beta blocker used to treat anxiety, hypertension and panic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 1 kg
Melting Range 162-165°C
Specific Rotation -1.0° to +1.0°
CAS Max % 1
Percent Purity 98 to 101.5%
Grade USP