Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprotein Convertase 9/PCSK9 Rabbit anti-Rat, DyLight 680, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290833FR
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Rat samples. It is validated for Sandwich ELISA Detection.Specifications
Proprotein Convertase 9/PCSK9 | |
Monoclonal | |
DyLight 680 | |
EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Proprotein Convertase 9/PCSK9 (NP_954862.2; Met1-Gln691). | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism | |
255738 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title