Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl Acetoacetate 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
1779-60-8
C7H12O3
144.17
MFCD00059405
DHGFMVMDBNLMKT-UHFFFAOYSA-N
Acetoacetic Acid Propyl Ester
74507
propyl 3-oxobutanoate
CCCOC(=O)CC(=O)C
1779-60-8
C7H12O3
144.17
MFCD00059405
DHGFMVMDBNLMKT-UHFFFAOYSA-N
Acetoacetic Acid Propyl Ester
74507
propyl 3-oxobutanoate
CCCOC(=O)CC(=O)C
Boiling Point 194°C
Color Colorless
Quantity 500mL
Formula Weight 144.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Propyl Acetoacetate

EINECSNumber : (2)-1475

TSCA : No