Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl propionate, 98+%, Acros Organics

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
Glass bottle
106-36-5
C6H12O2
116.16
MFCD00009373
MCSINKKTEDDPNK-UHFFFAOYSA-N
propyl propionate,n-propyl propionate,propanoic acid, propyl ester,n-propyl propanoate,propionic acid, propyl ester,n-propyl n-propionate,propylester kyseliny propionove,unii-g09trv00gk,propyl propionate natural,propionic acid n-propyl ester
7803
propyl propanoate
CCCOC(=O)CC
106-36-5
C6H12O2
116.16
MFCD00009373
MCSINKKTEDDPNK-UHFFFAOYSA-N
propyl propionate,n-propyl propionate,propanoic acid, propyl ester,n-propyl propanoate,propionic acid, propyl ester,n-propyl n-propionate,propylester kyseliny propionove,unii-g09trv00gk,propyl propionate natural,propionic acid n-propyl ester
7803
propyl propanoate
CCCOC(=O)CC
Refractive Index 1.3820 to 1.3940
Infrared Spectrum Authentic
Melting Point -76.0°C
Boiling Point 123.0°C
Color Colorless
Density 0.8810g/mL
Specific Gravity 0.881
Packaging Glass bottle
Quantity 1L
Flash Point 22°C
Assay Percent Range 98% min. (GC)
Linear Formula CH3CH2C(O)O(CH2)2CH3
Merck Index 15, 7979
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Propyl propionate
Show More Show Less

GHS H Statement
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a

GHS Signal Word: Danger

EINECSNumber : 203-389-7