Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
1L
5L
Packaging:
Glass Bottle
Plastic drum
107-10-8
C3H9N
59.11
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine,1-propanamine,1-propylamine,propanamine,n-propylamine,1-aminopropane,monopropylamine,mono-n-propylamine,propyl amine,n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN
107-10-8
C3H9N
59.11
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine,1-propanamine,1-propylamine,propanamine,n-propylamine,1-aminopropane,monopropylamine,mono-n-propylamine,propyl amine,n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN
Viscosity 0.4 mPa.s (20°C)
Packaging Plastic drum
Quantity 5L
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
Flash Point −30°C
Specific Gravity 0.719
Density 0.7190g/mL
Color Colorless to Yellow
Refractive Index 1.3870 to 1.3890
Melting Point -83.0°C
Boiling Point 48.0°C
Assay Percent Range 98%
Linear Formula CH3CH2CH2NH2
Beilstein 04,136
Fieser 13,164
Merck Index 15,7956
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether
Formula Weight 59.11
Physical Form Liquid
Percent Purity 98%
Grade Pure
Chemical Name or Material Propylamine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Refrigerator +4°C