Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylbenzene, ≥99% (GC), MilliporeSigma™ Supelco™

Propylbenzene belongs to the class of aromatic hydrocarbons, commonly used as a solvent in organic synthesis and precursor to synthesize intermediates such as 4-phenylbutyraldehyde and ethyl phenyl ketone.

$65.60 - $112.20

Specifications

CAS 103-65-1
Percent Purity ≥99% (GC)
Density 0.862 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula C6H5 CH2 CH2 CH3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9749
View Documents
MilliporeSigma Supelco
821185ML
5 mL
Each for $65.60
Add to cart
 
11-101-9015
View Documents
MilliporeSigma Supelco
8211810ML
10 mL
Each for $112.20
Add to cart
 
Description

Description

Featured Industries: Environmental
Specifications

Specifications

103-65-1
0.862 g/mL (at 25°C)
C6H5 CH2 CH2 CH3
≥99% (GC)
Analytical Standard
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit