Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propylene Carbonate, 99%, Alfa Aesar™
Click to view available options
Quantity:
250g
1000g
Description
Applications
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.
Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.
Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.
Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
Specifications
Refractive Index | 1.421 |
Flash Point | 132°C (270°F) |
Density | 1.204 |
Melting Point | °C to 49°C |
Boiling Point | 240°C to 243°C |
Quantity | 250g |
Assay Percent Range | 99% |
Solubility Information | Miscible with water,alcohol,ether,acetone,benzene,chloroform and ethyl acetate. |
Formula Weight | 102.09 |
Physical Form | Liquid |
Show More |
Safety and Handling
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 203-572-1
TSCA : Yes
Recommended Storage : Ambient temperatures