Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Carbonate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A155520I

Quantity 5kg

 View more versions of this product

Catalog No. AAA155520I

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.

Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.

Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
Description & Specifications

Chemical Identifiers

108-32-7
99%
102.09
Liquid
C4H6O3
Propylene Carbonate

Specifications

5kg
Liquid
C4H6O3
1.204
108-32-7
99%
102.09
Propylene Carbonate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit