Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Propylene Carbonate (Reagent), Fisher Chemical
Click to view available options
Quantity:
4L
Packaging:
Glass Bottle
Specifications
Identification | Pass Test |
Melting Point | -49°C |
pH | 5.5 to 7.5 |
Boiling Point | 240°C |
Quantity | 4L |
Density | 1.204g/cm³ |
Packaging | Glass Bottle |
Color | Colorless |
Assay Percent Range | ≥98 % |
Linear Formula | OCH(CH3)CH2COO |
Show More |