Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
19L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
57-55-6
C3H8O2
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol,propylene glycol,1,2-dihydroxypropane,2-hydroxypropanol,1,2-propylene glycol,methylethyl glycol,methylethylene glycol,monopropylene glycol,isopropylene glycol,dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O
57-55-6
C3H8O2
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol,propylene glycol,1,2-dihydroxypropane,2-hydroxypropanol,1,2-propylene glycol,methylethyl glycol,methylethylene glycol,monopropylene glycol,isopropylene glycol,dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 76.1
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Pail
Specific Gravity 1L = 1.04kg
Quantity 19L
Flash Point 99.00°C
Chemical Name or Material Propylene Glycol