Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
19L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail

Chemical Identifiers

57-55-6
C3H8O2
76.095
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1030
CHEBI:16997
CAS 57-55-6
Molecular Formula C3H8O2
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
SMILES CC(CO)O
IUPAC Name propane-1,2-diol
Molecular Weight (g/mol) 76.095
PubChem CID 1030
ChEBI CHEBI:16997
Formula Weight 76.1
Packaging Poly Pail
Specific Gravity 1L = 1.04kg
Quantity 19L
Flash Point 99.00°C
Chemical Name or Material Propylene Glycol
Grade Multi-Compendial/USP
Show More Show Less