Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol (USP/FCC), Fisher Chemical™

Click to view available options
Quantity:
1L
4L
20L
200L
Packaging:
Glass Bottle
Lined Steel Drum
Steel Drum
57-55-6
C3H8O2
76.095
MFCD00064272
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O
57-55-6
C3H8O2
76.095
MFCD00064272
DNIAPMSPPWPWGF-UHFFFAOYSA-N
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
1030
CHEBI:16997
propane-1,2-diol
CC(CO)O

Propylene Glycol (USP/FCC)

Acidity Pass Test (FCC/USP)
Density 2.62 (Air = 1.0)
pH 6 to 8
Color Undesignated
Melting Point -60°C
Boiling Point 187°C
Flash Point 99°C
Ignition Residue 0.007% max. (FCC/USP)
Identification Pass Test (FCC, USP)
Linear Formula CH3CHOHCH2OH
Packaging Lined Steel Drum
Quantity 20L
Specific Gravity 1.035 to 1.037 (FCC, USP)
Residual Solvents Meets Requirements (USP)
Formula Weight 76.1
Physical Form Viscous Liquid
Percent Purity ≥99.5% (USP)
Grade FCC/USP
Vapor Pressure 0.13mbar at 20°C
Water ≤0.2% (FCC, USP)
Chemical Name or Material Propylene Glycol
Show More Show Less

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:0
Flammability:1
Instability:0