Propylene Glycol, World/GMP, USEP/EP/JP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 369WORLDCS4L

Packaging Amber Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-101-440

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57-55-6
    Physical Form Liquid
    Molecular Formula C3H8O2
    Formula Weight 76.09
    Synonym 1, 2 Propanediol
    Chemical Name or Material Propylene Glycol

    Specifications

    Formula Weight 76.09
    Quantity 4L
    Boiling Point 188°C (370°F)
    Density 1.036g/mL at 25°C (77°F)
    Melting Point −60°C (−76°F)
    Flash Point 99°C (210°F)
    Color Colorless
    Physical Form Liquid
    Packaging Amber Glass Bottle
    Odor Mild
    Stability Stable under recommended storage conditions.
    Molecular Formula C3H8O2
    CAS 57-55-6
    Synonym 1, 2 Propanediol
    Chemical Name or Material Propylene Glycol

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit