Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
75-56-9
C3H6O
58.08
MFCD00005126
GOOHAUXETOMSMM-UHFFFAOYSA-N
propylene oxide,1,2-epoxypropane,methyloxirane,epoxypropane,propyleneoxide,propene oxide,oxirane, methyl,1,2-propylene oxide,propylene epoxide,2,3-epoxypropane
6378
CHEBI:38685
2-methyloxirane
CC1CO1
75-56-9
C3H6O
58.08
MFCD00005126
GOOHAUXETOMSMM-UHFFFAOYSA-N
propylene oxide,1,2-epoxypropane,methyloxirane,epoxypropane,propyleneoxide,propene oxide,oxirane, methyl,1,2-propylene oxide,propylene epoxide,2,3-epoxypropane
6378
CHEBI:38685
2-methyloxirane
CC1CO1
Viscosity 0.32 mPa.s (20°C)
Refractive Index 1.3650 to 1.3670
Water 0.02% max.
Specific Gravity 0.83
Melting Point -112.0°C
Packaging AcroSeal™ Glass Bottle
Density 0.8300g/mL
Quantity 100mL
Flash Point −37°C
Infrared Spectrum Authentic
Color Undesignated
Boiling Point 34.0°C
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.4% min. (GC)
Merck Index 15,7969
Solubility Information Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 58.08
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Pure
Chemical Name or Material (±)-Propylene oxide, Extra Pure
Show More Show Less

GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.

GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 200-879-2