Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Phenoxytol, Reagent, Spectrum™

Click to view available options
Quantity:
25 mL
125 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
770-35-4
C9H12O2
152.19
IBLKWZIFZMJLFL-UHFFFAOYNA-N
1-phenoxypropan-2-ol
CC(O)COC1=CC=CC=C1
770-35-4
C9H12O2
152.19
IBLKWZIFZMJLFL-UHFFFAOYNA-N
1-phenoxypropan-2-ol
CC(O)COC1=CC=CC=C1

PR132, 770-35-4, C9H12O2

Spectrum™ Propylene Phenoxytol, Reagent
Packaging Amber Glass Bottle
Quantity 125 mL
CAS Max % 1
Grade Reagent