Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylparaben Sodium, NF, 94-102%, Spectrum™

Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
35285-69-9
C10H11NaO3
202.19
IXMINYBUNCWGER-UHFFFAOYSA-M
sodium 4-(propoxycarbonyl)benzen-1-olate
[Na+].CCCOC(=O)C1=CC=C([O-])C=C1
35285-69-9
C10H11NaO3
202.19
IXMINYBUNCWGER-UHFFFAOYSA-M
sodium 4-(propoxycarbonyl)benzen-1-olate
[Na+].CCCOC(=O)C1=CC=C([O-])C=C1

P1457, 35285-69-9, C10H11NaO3

Spectrum™ Propylparaben Sodium, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Quantity 50 kg
Water 0.05
pH 9.5 to 10.5
CAS Max % 1
Percent Purity 94 to 102%
Grade NF