Learn More
Selleck Chemical LLCSupplier Diversity Partner Propylthiouracil 50mg 51-52-5 NSC 6498, NSC 70461  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S198850MG
Propylthiouracil (NSC 6498, NSC 70461) is a thyroperoxidase and 5'-deiodinase inhibitor, used to treat hyperthyroidism.
Catalog No. 50-695-2