Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyltriphenylphosphonium iodide, 99%, Acros Organics

Click to view available options
Quantity:
5g
25g
14350-50-6
C21H22IP
432.28
MPNQDJZRGAOBPW-UHFFFAOYSA-M
triphenyl propyl phosphonium iodide,propyltriphenylphosphonium iodide,triphenyl propyl phosphanium iodide,propyl triphenyl phosphonium iodide,n-propyl-triphenyl phosphonium iodide,phosphonium,triphenylpropyl-, iodide 1:1
10982905
triphenyl(propyl)phosphanium;iodide
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
14350-50-6
C21H22IP
432.28
MPNQDJZRGAOBPW-UHFFFAOYSA-M
triphenyl propyl phosphonium iodide,propyltriphenylphosphonium iodide,triphenyl propyl phosphanium iodide,propyl triphenyl phosphonium iodide,n-propyl-triphenyl phosphonium iodide,phosphonium,triphenylpropyl-, iodide 1:1
10982905
triphenyl(propyl)phosphanium;iodide
CCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
Melting Point 206.0°C to 209.0°C
Quantity 5g
Assay Percent Range 99%
Formula Weight 432.28
Percent Purity 99%
Chemical Name or Material Propyltriphenylphosphonium iodide

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS Signal Word: Warning