Learn More
ProSAPiP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 249361AP
Catalog No. 50-173-4628
Add to cart
Description
Description
Prosapip1 is a binding partner of the postsynaptic density (PSD) protein ProSAP2/Shank3, a major scaffolding protein in excitatory brain synapses. Prosapip1 is thought to bind ProSAP2/Shank3 through a PDZ domain in ProSAP2/Shank3 and link it in turn to the Spine-associated Rap-Gap (SPAR). Prosapip1 shares significant homology with another PSD protein, PSD-Zip70, with both coding for a Fez1 domain that can be found in a total of four related protein, forming a novel family of scaffolding molecules termed  ft.Fezzins ft.. Recent studies show that Prosapip1 can be found in endocrine cells in which it is localized primarily in the hormone-storing compartment of these cells, suggesting that Prosapip1 may have other functions in non-neuronal cells.
Specifications
Specifications
ProSAPiP1
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2AHG0, O60299
KIAA0552, ProSAP interacting protein 1, ProSAPiP1, ProSAPiP1 protein
Rabbit
IgG
150 μL
-20°C
Primary
241638, 9762
Western Blot
Unconjugated
LZTS3
Liquid
LZTS3
ProSAPiP1 Fusion Protein Ag20765
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse