Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PROSC Monoclonal Antibody (2G1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521300
Description
Peptide Sequence: MWRAGSMSAE LGVGCALRAV NERVQQAVAR RPRDLPAIQP RLVAVSKTKP ADMVIEAYGH GQRTFGENYV QELLEKASNP KILSLCPEIK WHFIGHLQKQ.
Pyridoxal 5'-phosphate (PLP)-binding protein, which may be involved in intracellular homeostatic regulation of pyridoxal 5'-phosphate (PLP), the active form of vitamin B6.
Specifications
PROSC | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
O94903 | |
PLPBP | |
PROSC (NP_009129.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA | |
2G1 | |
Unconjugated | |
PLPBP | |
1700024N20Rik; 2200002F22Rik; PLP homeostasis protein; PLPBP; Proline synthase co-transcribed bacterial homolog protein; proline synthetase co-transcribed; proline synthetase co-transcribed (bacterial homolog); proline synthetase co-transcribed bacterial homolog protein; proline synthetase co-transcribed homolog; proline synthetase cotranscribed homolog (bacterial); PROSC; Pyridoxal phosphate homeostasis protein; Pyridoxal phosphate-binding protein | |
Mouse | |
Affinity Chromatography | |
RUO | |
11212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction