Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROSC Mouse anti-Human, Clone: OTI3H4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA505130

Catalog No. 50-167-5674


Add to cart

Description

Description

Pyridoxal 5 ft.-phosphate (PLP)-binding protein, which may be involved in intracellular homeostatic regulation of pyridoxal 5 ft.-phosphate (PLP), the active form of vitamin B6.
Specifications

Specifications

PROSC
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O94903
EPVB6D, PROSC
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11212
Western Blot
OTI3H4
Unconjugated
PLPBP
Liquid
PLPBP
Full length human recombit protein of human PROSC produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit