Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 50 Coveralls with Elastic Wrist And Ankles

Made from microporous film, ProShield 50 garments help protect workers who face on-the-job exposure to a range of non-hazardous aerosols, light liquid splash and dry particles.

Manufacturer:  DuPont™ NB125SWHXL002500

 View more versions of this product

Catalog No. 19-065-502D


Add to cart

Alert:

Should not be used around heat, flame, sparks or potentially flammable or explosive environments.


Description

Description

  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Front zipper closure
  • Elastic wrist
  • Elastic ankles
  • Serged seams
  • Collar
  • White
  • Thickness: 10 mils
Specifications

Specifications

Elastic
Zipper front
White
Yes
Unisex
1.73 to 1.88m
88.90cm
Collared
Serged
40.75 to 44.25
71.75cm
10 mils
68.58cm
Safety
Lay down
Elastic
Janitorial, sanitation, and general industrial maintenance
5.22 to 6.16 ft.
35 in.
Composite of microporous film and nonwoven fabric.
Coveralls
X-Large
28.25 in.
Non-sterile
27 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit