Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 50 Coveralls with Hood and Boot

Made from microporous film, ProShield 50 garments help protect workers who face on-the-job exposure to a range of non-hazardous aerosols, light liquid splash and dry particles.

Manufacturer:  DuPont™ NB122SWHLG002500

 View more versions of this product

Catalog No. 19-065-501C


Add to cart

Alert:

Should not be used around heat, flame, sparks or potentially flammable or explosive environments.


Description

Description

  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Standard hood with elastic around the face
  • Attached skid-resistant boots
  • Front zipper closure
  • Elastic wrist
  • Serged seams
  • White
  • Thickness: 10 mils
Specifications

Specifications

Attached boots
Zipper front
Elastic
Janitorial, sanitation, and general industrial maintenance
5.41 to 5.75 ft.
Standard with elastic around the face
83.19cm
Hooded
Serged
37.25 to 40.75 in.
68.58cm
10 mils
64.135cm
Safety
White
Yes
Unisex
1.65 to 1.75m
32.75 in.
Composite of microporous film and nonwoven fabric.
Hooded coverall with attached boots
Large
27 in.
Non-sterile
25.25 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit