Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ PROSHIELD™ 6SFR Coverall with Standard Fit Hood, Attached Socks
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Description
DuPont™ ProShield® 6 SFR garments are cost-efficient and designed to be worn over primary flame resistant clothing.
- Keeps dirt and grime off of primary flame resistant garments
- Provides a barrier against non-hazardous particles and aerosols
Specifications
Size | Large |
Gender | Unisex |
Color | Blue |
Disposable | Single-use |
Ankle Style | Attached Socks, Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Seam Type | Serged |
Sterility | Non-sterile |
Resistance | Flame, Particle |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction