Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProShield™ 80 Coveralls
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Large
Medium
Small
X-Large
Description
- ProShield™ 80 coveralls provide a balance of comfort and protection against bloodborne pathogens.
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Garments feature a hydrophobic outer layer and soft inner layer, breathable fabric, and comfort fit design with gusset crotch.
- Can be considered a part of an overall PPE ensemble for occupations including emergency response, biohazard remediation, lab testing, and animal science.
- Bound seams exceed seam strength requirements as defined in ASTM F3352.
Specifications
Size | Medium |
Gender | Unisex |
Color | Blue |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Seam Type | Bound |
Sterility | Non-sterile |
Resistance | Bloodborne Pathogens |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction