Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProShield™ Disposable Lab Coat
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
Large
Medium
X-Large
Color:
Blue
White
Description
- These limited-use garments have been optimized for comfort, softness, and breathability
- Affordable and practical, ProShield™ garments can protect against non-hazardous dry particles and light liquid splash applications typically found in industrial situations
- For non-hazardous, light duty jobs, DuPont™ has the protection your workers need
Specifications
Size | 3X-Large |
Material | SMS |
Color | Blue |
Closure Type | Zipper |
Cuff Style | Elastic |
No. of Pockets | None |
For Use With (Application) | General Protection |
Length (Metric) | 43.5 |