Learn More
Thermo Scientific™ ProSpecT™ Campylobacter Microplate Assay Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Campylobacter specific antigen from human fecal samples.
Specifications
Description ProSpecT Campylobacter Microplate Assay
Storage Requirements 2°C to 8°C
View More Specs
Products 1
Catalog Number Mfr. No. Description Quantity Storage Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Storage Requirements Price Quantity & Availability  
R2476096 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R2476096
ProSpecT Campylobacter Microplate Assay 96 Tests/Kit 2°C to 8°C Pack of 96 for $1,951.95
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Rapidly detect Campylobacter specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Campylobacter Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Campylobacter. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Campylobacter is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Campylobacter provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ProSpecT Campylobacter Microplate Assay
2°C to 8°C