Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostaglandin dehydrogenase 1 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Supplier: Bioss BS6051R
Description
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.Specifications
Prostaglandin dehydrogenase 1 | |
Polyclonal | |
Unconjugated | |
PDHX | |
13G2AE2BH5; 15D3G9C11; 17A5E2H8; 4C11C10D12; 9H9AF5; Ac1164; AI481367; BOS_15094; BOS_26312; Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex; dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex (E3-binding protein) (proX); DLDBP; E3-binding protein; E3BP; Lipoyl-containing pyruvate dehydrogenase complex component X; LOC517402; OPDX; PDHA; PDHA1; Pdha-1; PDHAD; PDHCE1A; PDHE1-A type I; Pdhx; PDX1; PHE1A; proX; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase complex component X; pyruvate dehydrogenase complex, component X; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase complex, E3-binding protein subunit; pyruvate dehydrogenase complex, lipoyl-containing component X; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 component alpha subunit; Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate deh | |
Rabbit | |
Protein A | |
RUO | |
5160, 8050 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O00330, P08559 | |
PDHA1, PDHX | |
KLH conjugated synthetic peptide derived from human Prostaglandin dehydrogenase 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction