Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostasin/Prss8 Rabbit anti-Human, Clone: 107, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Prostasin/Prss8 |
---|---|
Clone | 107 |
Dilution | ELISA 1:1000-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Prostasin/Prss8 Monoclonal specifically detects Prostasin/Prss8 in Human samples. It is validated for ELISA.Specifications
Prostasin/Prss8 | |
ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
5652 | |
IgG | |
Protein A purified |
107 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, PROSTASIN, protease, serine, 8, Serine protease 8 | |
Recombinant Human Prostasin/Prss8 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title