Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS321R7

 View more versions of this product

Catalog No. MS321R7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Prostate Specific Acid Phosphatase (PSAP) Ab-1, Mouse Monoclonal Antibody.

Prostatic acid phosphatase (PSAP) is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase.

Host Species: Mouse

Clone: PASE/4LJ

Isotype: IgG1

Species Reactivity: Human. Does not react with dog, rabbit, and rat. Others not known.

Epitope: Not determined

Immunogen: Purified prostatic acid phosphatase (PSAP) from human seminal plasma

Molecular Weight: 52kDa

Positive Control: Normal prostate or prostate CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifications

Specifications

Prostate Specific Acid Phosphatase (PSAP) Ab-1
Monoclonal
Unconjugated
Prediluted
Purified prostatic acid phosphatase (PSAP) from human seminal plasma
7mL
Cancer and Tumor Biology
Primary
Human
Immunofluorescence, Immunohistochemistry (Paraffin)
PASE/4LJ
Ready-to-use for IHC (UVLP)
Murine
IgG1
IVD
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit