Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Prostatic Acid Phosphatase/ACPP Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796085PLATES

 View more versions of this product

Catalog No. NB010256


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Prostatic Acid Phosphatase/ACPP.
Specifications

Specifications

HRP
Antibody Pairs
156.25 pg/mL
55
RUO
Prostatic Acid Phosphatase/ACPP
Cell Cycle and Replication
Mouse
ACP3
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.4 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse Prostatic Acid Phosphatase/ACPP.
Sandwich ELISA
  • Standard: Each vial contains 810 ng of recombinant Mouse Prostatic Acid Phosphatase/ACPP.
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse Prostatic Acid Phosphatase/ACPP monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Prostatic Acid Phosphatase/ACPP monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    5 Plates
    156.25-10000 pg/mL
    acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit