Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostatic Acid Phosphatase/ACPP Mouse anti-Human, Allophycocyanin/Cy7, Clone: SPM312, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254475APCCY7

Catalog No. N254475APC7


Add to cart

Description

Description

Prostatic Acid Phosphatase/ACPP Monoclonal antibody specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Prostatic Acid Phosphatase/ACPP
SPM312
Flow Cytometry
Conjugated, Purified
ACPP
Prostatic acid phosphatase purified from human seminal plasma
Protein A or G purified
Cell Cycle and Replication
Primary
55.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit