Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prostatic Acid Phosphatase/ACPP Rabbit anti-Human, Clone: 60, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289722100UL
Description
Prostatic Acid Phosphatase/ACPP Monoclonal specifically detects Prostatic Acid Phosphatase/ACPP in Human samples. It is validated for ELISA.Specifications
Prostatic Acid Phosphatase/ACPP | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
60 | |
ELISA 1:5000-1:10000 | |
acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase | |
Recombinant Human Prostatic Acid Phosphatase/ACPP protein | |
100 μg | |
Cell Cycle and Replication | |
55 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction