Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prosurfactant Protein C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313377
This item is not returnable.
View return policy
Description
Prosurfactant Protein C Polyclonal antibody specifically detects Prosurfactant Protein C in Mouse, Rat samples. It is validated for Western BlotSpecifications
Prosurfactant Protein C | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
6440 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Prosurfactant Protein C. The exact sequence is proprietary. | |
100 μg | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction