Learn More
Prosurfactant Protein C Mouse anti-Human, DyLight 755, Clone: 5E6A9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237425IR
Catalog No. NBP237425IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Prosurfactant Protein C
5E6A9
Western Blot, ELISA
Purified
SFTPC
Purified recombinant fragment of human SFTPC (AA: 60-180 ) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C
Mouse
IgG1
0.1 ml
Primary
6440